Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_146152.3 | NP_666264.1 | 15 | 2988 | |
GenBank mRNA | AK052257.1 | - | 15 | 2989 | |
AK129200.1 | - | 15 | 2950 | ||
AK149978.1 | - | 15 | 2971 | ||
AK181621.1 | - | 2 | 119 | ||
AK191120.1 | - | 2 | 119 | ||
AK219089.1 | - | 2 | 119 | ||
BC031823.1 | - | 15 | 2387 | 4188768 |