Gene Transcripts

Gene Symbol
Cdk10
Entrez Gene ID
234854
Gene Name
cyclin-dependent kinase 10
Gene Aliases
BC017131
Chromosome Location
Chr. 8: 123224840 - 123232256 on Build GRCm38
UniGene ID
Mm.190984
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194444.2 NP_919426.2 5 356
NM_194446.2 NP_919428.1 5 394
GenBank mRNA AK133918.1 - 5 349
AK144801.1 - 5 366
AK170594.1 - 5 366
BC017131.1 - 5 331 4506458
BC022965.1 - 3 343
BC023736.1 - 5 375
BC052096.1 - 4 260
DQ288857.1 - 5 393

View Full Product Details