Gene Transcripts

Gene Symbol
Lsm14b
Entrez Gene ID
241846
Gene Name
LSM14 homolog B (SCD6, S. cerevisiae)
Gene Aliases
9430078B04, BC040823
Chromosome Location
Chr. 2: 180024902 - 180035466 on Build GRCm38
UniGene ID
Mm.21577
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177727.4 NP_808395.2 6 944
GenBank mRNA AK035040.1 - 6 973
AK162234.1 - 1 61
BC040823.1 - 6 944
BC099395.1 - 3 363 30933544
BY720115.1 - 1 59

View Full Product Details