Gene Transcripts

Gene Symbol
Dlc1
Entrez Gene ID
50768
Gene Name
deleted in liver cancer 1
Gene Aliases
A730069N07Rik, Arhgap7, HP, STARD12, dlc-1
Chromosome Location
Chr. 8: 36567739 - 36953165 on Build GRCm38
UniGene ID
Mm.125155
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001194940.2 NP_001181869.1 2 1040
GenBank mRNA AK043205.1 - 2 1022
AK045326.1 - 2 1035
AK053142.1 - 2 955
HM008381.1 - 1 593

View Full Product Details