Gene Transcripts

Gene Symbol
Olfr43
Entrez Gene ID
258706
Gene Name
olfactory receptor 43
Gene Aliases
IA7, MOR125-1, MOR125-5_p, Olfr403
Chromosome Location
Chr. 11: 74206198 - 74207289 on Build GRCm38
UniGene ID
Mm.453816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146711.2 NP_666922.2 1 471
GenBank mRNA BC125464.1 - 1 471
BC125468.1 - 1 471 40129983

View Full Product Details