Gene Transcripts

Gene Symbol
PSMD12
Entrez Gene ID
5718
Gene Name
proteasome 26S subunit, non-ATPase 12
Gene Aliases
Rpn5, p55
Chromosome Location
Chr. 17: 67337916 - 67366627 on Build GRCh38
UniGene ID
Hs.4295
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316341.1 NP_001303270.1 6 780
NM_002816.4 NP_002807.1 4 496
NM_174871.3 NP_777360.1 3 436
GenBank mRNA AB003103.1 - 4 431
AK091198.1 - 3 414
AK222679.1 - 4 413
AK308883.1 - 4 437
BC019062.2 - 4 427 4111477
BC065826.1 - 6 700 30386487
DC366628.1 - 4 506
KU178288.1 - 4 388
KU178289.1 - 4 388

View Full Product Details