Gene Transcripts

Gene Symbol
Colgalt2
Entrez Gene ID
269132
Gene Name
collagen beta(1-O)galactosyltransferase 2
Gene Aliases
AI427652, D730024P12, Glt25d2
Chromosome Location
Chr. 1: 152399867 - 152510695 on Build GRCm38
UniGene ID
Mm.23782
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177756.4 NP_808424.3 7 1283
GenBank mRNA AK052838.1 - 7 1284
AK076222.1 - 7 1025
AK085716.1 - 7 1323
AK134084.1 - 7 1264
BC068118.1 - 7 1229 5706069

View Full Product Details