Gene Transcripts

Gene Symbol
Pcnxl2
Entrez Gene ID
270109
Gene Name
pecanex-like 2 (Drosophila)
Gene Aliases
A830048P05, AU067631, E330039K12Rik, mKIAA0435
Chromosome Location
Chr. 8: 125751504 - 125898317 on Build GRCm38
UniGene ID
Mm.145535
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175561.4 NP_780770.2 24 4269
GenBank mRNA AK087907.1 - 9 1182
AK220342.1 - 16 1831
BC068235.1 - 13 2336

View Full Product Details