Gene Transcripts

Gene Symbol
Trav3n-3
Entrez Gene ID
100043282
Gene Name
T cell receptor alpha variable 3N-3
Gene Aliases
ENSMUSG00000076814, Gm10901
Chromosome Location
Chr. 14: 53283669 - 53284175 on Build GRCm38
UniGene ID
Mm.213248
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK041407.1 - 317
AK042313.1 - 310

Gene Symbol
Trav3-3
Entrez Gene ID
320898
Gene Name
T cell receptor alpha variable 3-3
Gene Aliases
A430107P09Rik, Gm13917, OTTMUSG00000015062
Chromosome Location
Chr. 14: 53666005 - 53666506 on Build GRCm38
UniGene ID
Mm.213248
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK134087.1 - 299

View Full Product Details