Gene Transcripts

Gene Symbol
Pramel4
Entrez Gene ID
347710
Gene Name
preferentially expressed antigen in melanoma like 4
Gene Aliases
C79430, D4Ertd792e
Chromosome Location
Chr. 4: 144059126 - 144069324 on Build GRCm38
UniGene ID
Mm.271697
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001319.3 NP_001001319.2 4 1547
GenBank mRNA AF490340.1 - 4 1431
AK145585.1 - 4 1548
AK163305.1 - 4 1406

Gene Symbol
Gm13102
Entrez Gene ID
100041077
Gene Name
predicted gene 13102
Gene Aliases
OTTMUSG00000010432
Chromosome Location
Chr. 4: 144099880 - 144110101 on Build GRCm38
UniGene ID
Mm.482724
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085419.1 NP_001078888.1 4 1546
GenBank mRNA BC132592.1 - 4 1547
BC132620.1 - 4 1547 40130897

View Full Product Details