Gene Transcripts

Gene Symbol
Pcdhac1
Entrez Gene ID
353236
Gene Name
protocadherin alpha subfamily C, 1
Gene Aliases
CNRc1
Chromosome Location
Chr. 18: 37090136 - 37187657 on Build GRCm38
UniGene ID
Mm.308500
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003671.1 NP_001003671.1 1 788
GenBank mRNA AY013768.1 - 1 788

View Full Product Details