Gene Transcripts

Gene Symbol
Rmdn2
Entrez Gene ID
381110
Gene Name
regulator of microtubule dynamics 2
Gene Aliases
AW061290, Fam82a1, RMD-2, mRMD-2
Chromosome Location
Chr. 17: 79578598 - 79685067 on Build GRCm38
UniGene ID
Mm.209801
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201361.2 NP_958749.1 2 382
GenBank mRNA AK034617.1 - 2 384
AK139382.1 - 2 601
BC024059.1 - 2 462 5067179

View Full Product Details