Gene Transcripts
- Gene Symbol
- Tmem181c-ps
- Entrez Gene ID
- 100040525
- Gene Name
- transmembrane protein 181C, pseudogene
- Gene Aliases
- 100040596, ENSMUSG00000079729, ENSMUSG00000079731, Tmem181d-ps
- Chromosome Location
- Chr. 17: 6610103 - 6620925 on Build GRCm38
- UniGene ID
- Mm.472852
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK045744.1 |
- |
2 |
596 |
|
- Gene Symbol
- Tmem181b-ps
- Entrez Gene ID
- 547127
- Gene Name
- transmembrane protein 181B, pseudogene
- Gene Aliases
- Tmem181b
- Chromosome Location
- Chr. 17: 6439002 - 6450994 on Build GRCm38
- UniGene ID
- Mm.297552
- Species
- Mus musculus
View Full Product Details