Gene Transcripts
- Gene Symbol
- RP9
- Entrez Gene ID
- 6100
- Gene Name
- retinitis pigmentosa 9 (autosomal dominant)
- Gene Aliases
- PAP-1, PAP1
- Chromosome Location
- Chr. 7: 33094797 - 33109390 on Build GRCh38
- UniGene ID
- Hs.326805
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_203288.1 |
NP_976033.1
|
1, 2 |
168 |
|
GenBank mRNA |
BC025928.1 |
- |
1, 2 |
168 |
|
View Full Product Details