Gene Transcripts

Gene Symbol
Psme2b
Entrez Gene ID
621823
Gene Name
protease (prosome, macropain) activator subunit 2B
Gene Aliases
PA28b-p, PA28b2, Psme2-like, Psme2b-ps
Chromosome Location
Chr. 11: 48945345 - 48946410 on Build GRCm38
UniGene ID
Mm.440151
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281472.1 NP_001268401.1 1 604
GenBank mRNA D87910.1 - 1 331

View Full Product Details