Gene Transcripts

Gene Symbol
Xlr3a
Entrez Gene ID
22445
Gene Name
X-linked lymphocyte-regulated 3A
Gene Aliases
Xlr3
Chromosome Location
Chr. X: 73086293 - 73097095 on Build GRCm38
UniGene ID
Mm.475098
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110784.1 NP_001104254.1 4 324
GenBank mRNA BC003812.1 - 4 270
BC093495.1 - 247 3593889
BC107255.1 - 233 40052405
BC107283.1 - 234 40052875
BC127145.1 - 234 40052876
BC127146.1 - 235 40052877
BQ885410.1 - 4 264
DV662513.1 - 4 272
U02599.1 - 3 217

Gene Symbol
Xlr3c
Entrez Gene ID
22446
Gene Name
X-linked lymphocyte-regulated 3C
Gene Aliases
Xlr3b
Chromosome Location
Chr. X: 73254538 - 73265571 on Build GRCm38
UniGene ID
Mm.336117
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011727.2 NP_035857.2 4 264
GenBank mRNA L22977.1 - 249
U02600.1 - 4 264

View Full Product Details