Gene Transcripts

Gene Symbol
S100A11
Entrez Gene ID
6282
Gene Name
S100 calcium binding protein A11
Gene Aliases
HEL-S-43, MLN70, S100C
Chromosome Location
Chr. 1: 152032506 - 152037035 on Build GRCh38
UniGene ID
Hs.417004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005620.1 NP_005611.1 2 247
GenBank mRNA AK312184.1 - 2 182
BC001410.2 - 2 207 3140092
BC014354.1 - 2 189 3685742
BT009912.1 - 1 127
D38583.1 - 2 247
D49355.1 - 2 214
D50374.1 - 2 143
EU794659.1 - 2 204
X80201.1 - 2 204

Gene Symbol
EIF1
Entrez Gene ID
10209
Gene Name
eukaryotic translation initiation factor 1
Gene Aliases
A121, EIF-1, EIF1A, ISO1, SUI1
Chromosome Location
Chr. 17: 41688875 - 41691646 on Build GRCh38
UniGene ID
Hs.150580
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AA159721.1 - 390

View Full Product Details