Gene Transcripts
- Gene Symbol
- LOC665506
- Entrez Gene ID
- 665506
- Gene Name
- T-cell receptor beta-2 chain C region-like
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.333026
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X00619.1 |
- |
|
388 |
|
- Gene Symbol
- Tcrb-J
- Entrez Gene ID
- 21580
- Gene Name
- T cell receptor beta, joining region
- Gene Aliases
- 5830405F06Rik
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.333026
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M11456.1 |
- |
4 |
481 |
|
X00438.1 |
- |
4 |
485 |
|
- Gene Symbol
- Trbv1
- Entrez Gene ID
- 621968
- Gene Name
- T cell receptor beta, variable 1
- Gene Aliases
- Gm6273, TCRBV2S1, Tcrb-V2
- Chromosome Location
- Chr. 6: 40891296 - 40891885 on Build GRCm38
- UniGene ID
- Mm.333009
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK138971.1 |
- |
|
535 |
|
- Gene Symbol
- Trbv13-2
- Entrez Gene ID
- 100124675
- Gene Name
- T cell receptor beta, variable 13-2
- Gene Aliases
- TCRBV8S2, Tcrb, Tcrb-V8.2
- Chromosome Location
- Chr. 6: 41121396 - 41121832 on Build GRCm38
- UniGene ID
- Mm.123831
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
L37870.1 |
- |
|
418 |
|
L37872.1 |
- |
|
437 |
|
View Full Product Details