Gene Transcripts
- Gene Symbol
- Nkx1-1
- Entrez Gene ID
- 672284
- Gene Name
- NK1 transcription factor related, locus 1 (Drosophila)
- Gene Aliases
- Nkx-1.1, Sax2
- Chromosome Location
- Chr. 5: 33430734 - 33434090 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011320.1 |
NP_035450.1
|
1 |
284 |
|
View Full Product Details