Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081679.1 | NP_001075148.1 | 4 | 741 | |
NM_001282088.1 | NP_001269017.1 | 4 | 750 | ||
GenBank mRNA | AK158024.1 | - | 751 | ||
DQ854743.1 | - | 4 | 741 | ||
EU627091.1 | - | 4 | 750 |