Gene Transcripts
- Gene Symbol
- Slc9a1
- Entrez Gene ID
- 24782
- Gene Name
- solute carrier family 9, subfamily A (NHE1, cation proton antiporter 1), member 1
- Gene Aliases
- Nhe1
- Chromosome Location
- Chr. 5: 151573122 - 151626360 on Build Rnor_6.0
- UniGene ID
- Rn.5025
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012652.1 |
NP_036784.1
|
9, 10 |
2716 |
|
GenBank mRNA |
M85299.1 |
- |
9, 10 |
2716 |
|
View Full Product Details