Gene Transcripts

Gene Symbol
Thrsp
Entrez Gene ID
25357
Gene Name
thyroid hormone responsive
Gene Aliases
Lpgp, S14, SPOT14
Chromosome Location
Chr. 1: 162381251 - 162385575 on Build Rnor_6.0
UniGene ID
Rn.81140
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012703.2 NP_036835.2 1 292
GenBank mRNA BC089061.1 - 1 292
FQ214269.1 - 1 303

View Full Product Details