Gene Transcripts

Gene Symbol
Dync1i1
Entrez Gene ID
29564
Gene Name
dynein cytoplasmic 1 intermediate chain 1
Gene Aliases
Dnci1, Dncic1
Chromosome Location
Chr. 4: 30807704 - 31121095 on Build Rnor_6.0
UniGene ID
Rn.11273
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019234.1 NP_062107.1 8 867
XM_006236026.2 XP_006236088.1 8 994
GenBank mRNA X66845.1 - 8 867

View Full Product Details