Gene Transcripts

Gene Symbol
SIGLEC1
Entrez Gene ID
6614
Gene Name
sialic acid binding Ig like lectin 1
Gene Aliases
CD169, SIGLEC-1, SN, dJ1009E24.1
Chromosome Location
Chr. 20: 3686970 - 3712558 on Build GRCh38
UniGene ID
Hs.31869
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023068.3 NP_075556.1 20, 21 5068
XM_006723610.3 XP_006723673.1 21, 22 5263
XM_011529324.2 XP_011527626.1 21, 22 5260
XM_011529325.2 XP_011527627.1 21, 22 5266
XM_011529326.2 XP_011527628.1 21, 22 5262
XM_011529327.2 XP_011527629.1 20, 21 5196
XM_011529328.2 XP_011527630.1 21, 22 5208
XM_011529330.1 XP_011527632.1 19, 20 6191
XM_011529331.1 XP_011527633.1 20, 21 6367
XM_011529332.1 XP_011527634.1 19, 20 4653
GenBank mRNA AF230073.1 - 20, 21 5068
AK024462.1 - 12, 13 2872
AL833688.1 - 5, 6 803

View Full Product Details