Gene Transcripts

Gene Symbol
SOD3
Entrez Gene ID
6649
Gene Name
superoxide dismutase 3, extracellular
Gene Aliases
EC-SOD
Chromosome Location
Chr. 4: 24795463 - 24800845 on Build GRCh38
UniGene ID
Hs.2420
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003102.2 NP_003093.2 2 1433
GenBank mRNA BC014418.1 - 2 1298
J02947.1 - 2 1297

View Full Product Details