Gene Transcripts

Gene Symbol
Tsnax
Entrez Gene ID
64028
Gene Name
translin-associated factor X
Gene Aliases
Trax
Chromosome Location
Chr. 19: 57771539 - 57785567 on Build Rnor_6.0
UniGene ID
Rn.2315
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022262.3 NP_071598.1 3 238
GenBank mRNA AF262357.1 - 3 309
BC081715.1 - 3 238 7130295

View Full Product Details