Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003126.2 | NP_003117.2 | 15 | 2148 | |
XM_011509916.2 | XP_011508218.1 | 16 | 2026 | ||
XM_011509917.2 | XP_011508219.1 | 15 | 2062 | ||
XM_011509918.2 | XP_011508220.1 | 15 | 2062 | ||
XM_011509919.2 | XP_011508221.1 | 15 | 2064 | ||
GenBank mRNA | M61877.1 | - | 15 | 2135 |