Gene Transcripts

Gene Symbol
SRM
Entrez Gene ID
6723
Gene Name
spermidine synthase
Gene Aliases
PAPT, SPDSY, SPS1, SRML1
Chromosome Location
Chr. 1: 11054592 - 11060053 on Build GRCh38
UniGene ID
Hs.76244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003132.2 NP_003123.2 4 552
XM_017002179.1 XP_016857668.1 4 571
GenBank mRNA BC000309.2 - 4 518
BC033106.1 - 4 484 5420683
M34338.1 - 4 543

View Full Product Details