Gene Transcripts

Gene Symbol
Scamp2
Entrez Gene ID
65168
Gene Name
secretory carrier membrane protein 2
Gene Aliases
Chromosome Location
Chr. 8: 62341613 - 62368215 on Build Rnor_6.0
UniGene ID
Rn.3752
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023955.1 NP_076445.1 4, 5 432
GenBank mRNA AF295405.1 - 3, 4 276
BC085862.1 - 4, 5 432 7190371

View Full Product Details