Gene Transcripts

Gene Symbol
Gcs1
Entrez Gene ID
78947
Gene Name
glucosidase 1
Gene Aliases
Mogs
Chromosome Location
Chr. 4: 113948328 - 113951731 on Build Rnor_6.0
UniGene ID
Rn.22161
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031749.2 NP_113937.1 4 2225
GenBank mRNA AF087431.1 - 4 2226

Gene Symbol
LOC103692171
Entrez Gene ID
103692171
Gene Name
mannosyl-oligosaccharide glucosidase
Gene Aliases
Chromosome Location
Chr. 4: 114813988 - 114817463 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008763086.1 XP_008761308.1 2293

View Full Product Details