Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031102.2 | NP_112364.1 | 3 | 158 | |
XM_006229161.2 | XP_006229223.1 | 3 | 639 | ||
GenBank mRNA | BC084727.1 | - | 2 | 150 | |
CB577548.1 | - | 3 | 158 | ||
DQ403026.1 | - | 2 | 74 | ||
FQ211765.1 | - | 3 | 176 | ||
FQ221110.1 | - | 3 | 176 | ||
FQ222651.1 | - | 3 | 176 | ||
FQ228615.1 | - | 3 | 179 | ||
M20156.1 | - | 2 | 144 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008773122.1 | XP_008771344.1 | 1 | 141 |