Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031102.2 | NP_112364.1 | 4 | 240 | |
XM_006229161.2 | XP_006229223.1 | 4 | 721 | ||
GenBank mRNA | BC084727.1 | - | 3 | 232 | |
CB577548.1 | - | 4 | 240 | ||
DQ403026.1 | - | 3 | 156 | ||
FQ211765.1 | - | 4 | 258 | ||
FQ221110.1 | - | 4 | 258 | ||
FQ222651.1 | - | 4 | 258 | ||
FQ228615.1 | - | 4 | 261 | ||
M20156.1 | - | 3 | 226 |