Gene Transcripts

Gene Symbol
Synrg
Entrez Gene ID
84479
Gene Name
synergin, gamma
Gene Aliases
Ap1gbp1, Syng
Chromosome Location
Chr. 10: 71278698 - 71357791 on Build Rnor_6.0
UniGene ID
Rn.122513
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053419.1 NP_445871.1 8 914
GenBank mRNA EF121979.1 - 7 611
EF121980.1 - 7 611
EF121981.1 - 8 914

View Full Product Details