Gene Transcripts

Gene Symbol
TBL1X
Entrez Gene ID
6907
Gene Name
transducin (beta)-like 1X-linked
Gene Aliases
EBI, SMAP55, TBL1
Chromosome Location
Chr. X: 9463295 - 9719740 on Build GRCh38
UniGene ID
Hs.495656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001139466.1 NP_001132938.1 11 1376
NM_001139467.1 NP_001132939.1 10 1231
NM_001139468.1 NP_001132940.1 11 1366
NM_005647.3 NP_005638.1 11 1495
XM_011545571.2 XP_011543873.1 10 1217
GenBank mRNA AB209608.1 - 6 1449
AK289409.1 - 10 1231
AK290962.1 - 11 1359
BC032708.1 - 11 1342 5496758
BC052304.1 - 11 1495 6378852
Y12781.1 - 11 1654

View Full Product Details