Gene Transcripts

Gene Symbol
DYNLT3
Entrez Gene ID
6990
Gene Name
dynein light chain Tctex-type 3
Gene Aliases
RP3, TCTE1L, TCTEX1L
Chromosome Location
Chr. X: 37838836 - 37847636 on Build GRCh38
UniGene ID
Hs.446392
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006520.2 NP_006511.1 3 257
GenBank mRNA AK311851.1 - 3 192
BC000968.2 - 3 185 3448623
CR450320.1 - 3 131
CR542213.1 - 3 131
DA342302.1 - 3 248
DB492712.1 - 3 258
U02556.1 - 3 199

View Full Product Details