Gene Transcripts

Gene Symbol
Pvrl1
Entrez Gene ID
192183
Gene Name
poliovirus receptor-related 1
Gene Aliases
HveC, nectin-1
Chromosome Location
Chr. 8: 48094233 - 48157315 on Build Rnor_6.0
UniGene ID
Rn.156563
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006242963.2 XP_006243025.1 6 1980
XM_236210.8 XP_236210.3 6 1870

View Full Product Details