Gene Transcripts
- Gene Symbol
- TGFBI
- Entrez Gene ID
- 7045
- Gene Name
- transforming growth factor beta induced
- Gene Aliases
- BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, LCD1
- Chromosome Location
- Chr. 5: 136028895 - 136063818 on Build GRCh38
- UniGene ID
- Hs.369397
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000358.2 |
NP_000349.1
|
5, 6 |
786 |
|
GenBank mRNA |
AK093916.1 |
- |
5, 6 |
641 |
|
AK094055.1 |
- |
6, 7 |
717 |
|
AK094581.1 |
- |
5, 6 |
730 |
|
AK222833.1 |
- |
5, 6 |
694 |
|
AK303643.1 |
- |
5, 6 |
1018 |
|
BC000097.2 |
- |
5, 6 |
679 |
3506051 |
BC004972.1 |
- |
5, 6 |
674 |
2958878 |
BT009820.1 |
- |
5, 6 |
625 |
|
KU178357.1 |
- |
5, 6 |
625 |
|
M77349.1 |
- |
5, 6 |
672 |
|
View Full Product Details