Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001323316.1 | NP_001310245.1 | 11 | 1596 | |
NM_001323317.1 | NP_001310246.1 | 9 | 1370 | ||
NM_001323318.1 | NP_001310247.1 | 9 | 1433 | ||
NM_004613.3 | NP_004604.2 | 10 | 1613 | ||
NM_198951.2 | NP_945189.1 | 10 | 1613 | ||
XM_011529028.1 | XP_011527330.1 | 11 | 1558 | ||
GenBank mRNA | AK058031.1 | - | 10 | 1381 | |
AK291714.1 | - | 10 | 1454 | ||
AK295775.1 | - | 9 | 1211 | ||
AK300292.1 | - | 9 | 1274 | ||
AK314618.1 | - | 10 | 1454 | ||
AL512703.1 | - | 1 | 2182 | ||
AY675221.1 | - | 9 | 1382 | ||
BC003551.1 | - | 10 | 1466 | 3544757 | |
BQ938957.1 | - | 2 | 174 | ||
EU794665.1 | - | 10 | 1482 | ||
M55153.1 | - | 10 | 1517 | ||
M98478.1 | - | 10 | 1514 |