Gene Transcripts

Gene Symbol
Dpy30
Entrez Gene ID
286897
Gene Name
dpy-30 homolog (C. elegans)
Gene Aliases
Aip1
Chromosome Location
Chr. 6: 22296128 - 22316894 on Build Rnor_6.0
UniGene ID
Rn.3297
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170545.1 NP_001164016.1 4 316
NM_173117.2 NP_775140.2 4 355
GenBank mRNA AY129401.1 - 4 355
FM121127.1 - 4 322

View Full Product Details