Gene Transcripts
- Gene Symbol
- Ptx4
- Entrez Gene ID
- 287130
- Gene Name
- pentraxin 4, long
- Gene Aliases
- RGD1311190
- Chromosome Location
- Chr. 10: 14481044 - 14486277 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_220237.4 |
XP_220237.4
|
3 |
1314 |
|
View Full Product Details