Gene Transcripts

Gene Symbol
Nt5m
Entrez Gene ID
287368
Gene Name
5',3'-nucleotidase, mitochondrial
Gene Aliases
Chromosome Location
Chr. 10: 46216493 - 46244476 on Build Rnor_6.0
UniGene ID
Rn.2189
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105785.1 NP_001099255.1 4 443
XM_006246460.2 XP_006246522.1 4 890
GenBank mRNA BC167058.1 - 4 499

View Full Product Details