Gene Transcripts

Gene Symbol
Nmnat2
Entrez Gene ID
289095
Gene Name
nicotinamide nucleotide adenylyltransferase 2
Gene Aliases
Chromosome Location
Chr. 13: 70379346 - 70559311 on Build Rnor_6.0
UniGene ID
Rn.145458
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048042.1 NP_001041507.1 9 869
XM_008769614.1 XP_008767836.1 9 727
GenBank mRNA DQ022370.1 - 9 869

View Full Product Details