Gene Transcripts
- Gene Symbol
- Prg4
- Entrez Gene ID
- 289104
- Gene Name
- proteoglycan 4
- Gene Aliases
- Chromosome Location
- Chr. 13: 67672588 - 67688902 on Build Rnor_6.0
- UniGene ID
- Rn.198420
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC158740.1 |
- |
3 |
718 |
|
View Full Product Details