Gene Transcripts

Gene Symbol
Mrc1
Entrez Gene ID
291327
Gene Name
mannose receptor, C type 1
Gene Aliases
Chromosome Location
Chr. 17: 81352700 - 81433743 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106123.2 NP_001099593.2 23 3326

View Full Product Details