Gene Transcripts

Gene Symbol
Sprtn
Entrez Gene ID
292101
Gene Name
SprT-like N-terminal domain
Gene Aliases
RGD1559496
Chromosome Location
Chr. 19: 57649900 - 57657158 on Build Rnor_6.0
UniGene ID
Rn.144231
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006255808.2 XP_006255870.1 3 622
XM_006255809.2 XP_006255871.1 3 379
XM_006255810.2 XP_006255872.1 3 405

View Full Product Details