Gene Transcripts

Gene Symbol
Sult2b1
Entrez Gene ID
292915
Gene Name
sulfotransferase family, cytosolic, 2B, member 1
Gene Aliases
ST2B1
Chromosome Location
Chr. 1: 101712254 - 101773392 on Build Rnor_6.0
UniGene ID
Rn.100005
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039665.1 NP_001034754.1 3 560
XM_006229031.2 XP_006229093.1 3 461
XM_006229032.2 XP_006229094.1 3 426
XM_006229033.2 XP_006229095.1 3 792
XM_008759338.1 XP_008757560.1 3 389
GenBank mRNA AY827147.1 - 3 560
AY827148.1 - 3 429
BC127438.1 - 1234

View Full Product Details