Gene Transcripts

Gene Symbol
Olr20
Entrez Gene ID
293083
Gene Name
olfactory receptor 20
Gene Aliases
Chromosome Location
Chr. 1: 149963547 - 149964539 on Build Rnor_6.0
UniGene ID
Rn.141948
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000118.1 NP_001000118.1 1 910

Gene Symbol
LOC100911035
Entrez Gene ID
100911035
Gene Name
olfactory receptor 14C36-like
Gene Aliases
Chromosome Location
Chr. 1|NW_007906107: 49780 - 50772 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003752172.2 XP_003752220.2 1 910

View Full Product Details