Gene Transcripts

Gene Symbol
Micalcl
Entrez Gene ID
293180
Gene Name
MICAL C-terminal like
Gene Aliases
RSB-11-77
Chromosome Location
Chr. 1: 177193735 - 177232674 on Build Rnor_6.0
UniGene ID
Rn.30134
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182669.2 NP_872610.2 9 2012
GenBank mRNA AY149342.1 - 6 1452
BC098647.1 - 9 2012 7112444
BC100137.1 - 6 1466 7457561

View Full Product Details