Gene Transcripts
- Gene Symbol
- UROS
- Entrez Gene ID
- 7390
- Gene Name
- uroporphyrinogen III synthase
- Gene Aliases
- UROIIIS
- Chromosome Location
- Chr. 10: 125784980 - 125823268 on Build GRCh38
- UniGene ID
- Hs.501376
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000375.2 |
NP_000366.1
|
5, 6 |
582 |
|
NM_001324036.1 |
NP_001310965.1
|
5, 6 |
582 |
|
NM_001324037.1 |
NP_001310966.1
|
5, 6 |
582 |
|
NM_001324038.1 |
NP_001310967.1
|
5, 6 |
582 |
|
NM_001324039.1 |
NP_001310968.1
|
5, 6 |
582 |
|
NR_136675.1 |
|
3, 4 |
401 |
|
NR_136676.1 |
|
5, 6 |
582 |
|
NR_136677.1 |
|
5, 6 |
582 |
|
NR_136678.1 |
|
2, 3 |
312 |
|
XM_005270140.4 |
XP_005270197.1
|
5, 6 |
424 |
|
XM_011540127.1 |
XP_011538429.1
|
5, 6 |
424 |
|
XM_017016610.1 |
XP_016872099.1
|
5, 6 |
424 |
|
XM_017016611.1 |
XP_016872100.1
|
5, 6 |
380 |
|
XM_017016612.1 |
XP_016872101.1
|
5, 6 |
424 |
|
GenBank mRNA |
AF230665.1 |
- |
4, 5 |
449 |
|
AK314896.1 |
- |
5, 6 |
439 |
|
BC002573.2 |
- |
5, 6 |
541 |
3160463 |
BI668388.1 |
- |
2, 3 |
252 |
|
J03824.1 |
- |
5, 6 |
512 |
|
KU178377.1 |
- |
4, 5 |
316 |
|
KU178378.1 |
- |
4, 5 |
316 |
|
View Full Product Details