Gene Transcripts

Gene Symbol
VASP
Entrez Gene ID
7408
Gene Name
vasodilator-stimulated phosphoprotein
Gene Aliases
Chromosome Location
Chr. 19: 45507430 - 45526989 on Build GRCh38
UniGene ID
Hs.515469
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003370.3 NP_003361.1 11 1366
XM_005259199.1 XP_005259256.1 11 1351
XM_005259200.1 XP_005259257.1 11 1351
XM_017027200.1 XP_016882689.1 11 1348
GenBank mRNA AK314812.1 - 11 1333
AL556269.3 - 9 841
BC026019.1 - 11 1303 3345963
BC038224.1 - 11 1284 5210128
KU178381.1 - 10 1024
KU178382.1 - 11 868
KU178383.1 - 11 865
KU178384.1 - 10 748
KU178385.1 - 8 573
Z46389.1 - 11 1278

View Full Product Details